Learn More
Medchemexpress LLC Jpm-oet | 262381-84-0 | 99.0% | 392.45 g/mol | C20H28N2O6 | 10 MG
SDP
Catalog No. 5000247915
Encompass_Preferred
JPM-OEt is a broad-spectrum cysteine cathepsin inhibitor that binds covalently in the active site and irreversibly inhibits cysteine cathepsins. It has demonstrated antitumor activity in preclinical studies and is supplied as a solid for research use.
  • Covalent, irreversible inhibitor of cysteine cathepsins.
  • Demonstrated antitumor activity in preclinical models.
  • High purity solid (98.99%), white to off-white appearance.
  • High solubility in DMSO; in vivo formulation protocols available.
  • Storage guidance provided for solid and solution forms.
Catalog No. 50-002-47915 Supplier Medchemexpress LLC Supplier No. HY10208710MG
JPM-OEt is a broad-spectrum cysteine cathepsin inhibitor that binds covalently in the active site and irreversibly inhibits cysteine cathepsins. It has demonstrated antitumor activity in preclinical studies and is supplied as a solid for research use.
  • Covalent, irreversible inhibitor of cysteine cathepsins.
  • Demonstrated antitumor activity in preclinical models.
  • High purity solid (98.99%), white to off-white appearance.
  • High solubility in DMSO; in vivo formulation protocols available.
  • Storage guidance provided for solid and solution forms.