Learn More
Medchemexpress LLC Jpm-oet | 262381-84-0 | 99.0% | 392.45 g/mol | C20H28N2O6 | 100 MG
SDP
Catalog No. 5000245534
Encompass_Preferred
JPM-OEt is a covalent, irreversible inhibitor of the cysteine cathepsin family used in biochemical and in vivo research to study protease-dependent processes and tumor models.
  • Broad-spectrum cysteine cathepsin inhibitor that binds covalently and irreversibly.
  • Demonstrated antitumor activity in preclinical studies.
  • High analytical purity (98.99%).
  • Supplied as a white to off-white solid, available in multiple pack sizes.
  • DMSO solubility approximately 125 mg/mL; suitable in vivo formulations reported.
  • Store protected from light; solvent storage recommendations range from -80°C to 4°C.
Catalog No. 50-002-45534 Supplier Medchemexpress LLC Supplier No. HY102087100MG
JPM-OEt is a covalent, irreversible inhibitor of the cysteine cathepsin family used in biochemical and in vivo research to study protease-dependent processes and tumor models.
  • Broad-spectrum cysteine cathepsin inhibitor that binds covalently and irreversibly.
  • Demonstrated antitumor activity in preclinical studies.
  • High analytical purity (98.99%).
  • Supplied as a white to off-white solid, available in multiple pack sizes.
  • DMSO solubility approximately 125 mg/mL; suitable in vivo formulations reported.
  • Store protected from light; solvent storage recommendations range from -80°C to 4°C.