Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JPM-OEt | 262381-84-0 | MFCD16660940 | 99.0% | 392.45 g·mol⁻¹ | C20H28N2O6 | 50 MG
SDP

Catalog No. 5000245559
Encompass_Preferred

JPM-OEt is a covalent, irreversible broad-spectrum inhibitor of cysteine cathepsins used in biochemical and cell-based research; it has reported antitumor activity.

  • Broad spectrum cysteine cathepsin inhibitor; covalent, irreversible binding
  • Chemical formula C20H28N2O6; molecular weight 392.45 g·mol⁻¹
  • Supplied as a white to off-white solid
  • Reported purity 98.99% (≈99.0%)
  • Typical applications: biochemical assays, cell-based studies, preclinical research
  • Storage: store at 4°C protected from light; in solvent, -80°C up to 6 months, -20°C up to 1 month
  • CAS number 262381-84-0

Catalog No. 50-002-45559 Supplier Medchemexpress LLC Supplier No. HY10208750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JPM-OEt is a covalent, irreversible broad-spectrum inhibitor of cysteine cathepsins used in biochemical and cell-based research; it has reported antitumor activity.

  • Broad spectrum cysteine cathepsin inhibitor; covalent, irreversible binding
  • Chemical formula C20H28N2O6; molecular weight 392.45 g·mol⁻¹
  • Supplied as a white to off-white solid
  • Reported purity 98.99% (≈99.0%)
  • Typical applications: biochemical assays, cell-based studies, preclinical research
  • Storage: store at 4°C protected from light; in solvent, -80°C up to 6 months, -20°C up to 1 month
  • CAS number 262381-84-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.