Learn More
Medchemexpress LLC HY-13030 500mg , (+)-JQ-1 CAS:1268524-70-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13030500MG
Encompass_Preferred
HY-13030 500mg (+)-JQ-1 CAS: 1268524-70-4 (+)-JQ-1 (JQ1) is a potent, specific, and reversible BET bromodomain inhibitor, with IC50s of 77 and 33 nM for the first and second bromodomain (BRD4(1/2)). (+)-JQ-1 also activates autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9078