Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-JQ-1-aldehyde | 2634778-37-1 | 98.1% | C19H17ClN4OS | 10MG
SDP

Catalog No. 5000261504
Encompass_Preferred

(+)-JQ-1-aldehyde is the aldehyde derivative of the BET inhibitor JQ1 and is used as a synthetic precursor for PROTACs and other derivatives that target BET bromodomains. It is supplied as a light yellow solid with documented solubility and storage recommendations for research applications.

  • Used as a synthetic precursor for PROTACs and derivatives.
  • High purity (98.1%).
  • Light yellow solid appearance.
  • Chemical formula C19H17ClN4OS and molecular weight 384.88 g/mol.
  • Soluble in DMSO (100 mg/mL); sonication may be required.
  • Example in vivo formulations achievable at ≥2.5 mg/mL in mixed vehicles.
  • Store solid sealed at 4°C; in solution store at -80°C for long term.

Catalog No. 50-002-61504 Supplier Medchemexpress LLC Supplier No. HY131633A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-JQ-1-aldehyde is the aldehyde derivative of the BET inhibitor JQ1 and is used as a synthetic precursor for PROTACs and other derivatives that target BET bromodomains. It is supplied as a light yellow solid with documented solubility and storage recommendations for research applications.

  • Used as a synthetic precursor for PROTACs and derivatives.
  • High purity (98.1%).
  • Light yellow solid appearance.
  • Chemical formula C19H17ClN4OS and molecular weight 384.88 g/mol.
  • Soluble in DMSO (100 mg/mL); sonication may be required.
  • Example in vivo formulations achievable at ≥2.5 mg/mL in mixed vehicles.
  • Store solid sealed at 4°C; in solution store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.