Learn More
Medchemexpress LLC HY-131633A 5mg Medchemexpress, (+)-JQ-1-aldehyde CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131633A5MG
Encompass_Preferred
Medchemexpress, HY-131633A 5mg (+)-JQ-1-aldehyde CAS: (+)-JQ-1-aldehyde is the aldehyde form of (+)-JQ1. (+)-JQ-1-aldehyde can be uesd as a precursor to synthesize PROTACs, which targets BET bromodomains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1290