Learn More
Medchemexpress LLC JQ-1 (carboxylic acid) | 202592-23-2 | C19H17ClN4O2S | 100 MG
SDP
Catalog No. 502029368
Encompass_Preferred
JQ-1 carboxylic acid, a derivative of (+)-JQ-1, is a potent inhibitor of BET bromodomain. This compound can be utilized in the synthesis of PROTACs, enabling the targeted degradation of BRD4.
  • Target: Epigenetic reader domain, PD-1/PD-L1
  • Research areas: Cancer
  • Mechanism of action: Potent BET bromodomain inhibitor; can be used to synthesize PROTAC, which can target the degradation of BRD4.
Catalog No. 50-202-9368 Supplier Medchemexpress LLC Supplier No. HY78695100MG
JQ-1 carboxylic acid, a derivative of (+)-JQ-1, is a potent inhibitor of BET bromodomain. This compound can be utilized in the synthesis of PROTACs, enabling the targeted degradation of BRD4.
  • Target: Epigenetic reader domain, PD-1/PD-L1
  • Research areas: Cancer
  • Mechanism of action: Potent BET bromodomain inhibitor; can be used to synthesize PROTAC, which can target the degradation of BRD4.