Learn More
Medchemexpress LLC JQ-1 (carboxylic acid) | 202592-23-2 | 99.7% | 400.88 | 50 MG
SDP
Catalog No. 5000364274
Encompass_Preferred
JQ-1 carboxylic acid is a potent BET bromodomain inhibitor, derived from (+)-JQ-1. It is utilized for synthesizing PROTACs that target the degradation of BRD4. This compound is an off-white to yellow solid with a molecular weight of 400.88 and the chemical formula C19H17ClN4O2S.
  • Potent BET bromodomain inhibitor.
  • Used to synthesize PROTAC, targeting BRD4 degradation.
  • Soluble in DMSO (100 mg/mL).
  • Downregulates PD-L1 expression on B16F10 cells in vitro.
  • Applicable in various disease areas including cancer, and signaling pathways such as epigenetics and immunology/inflammation.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
Catalog No. 50-003-64274 Supplier Medchemexpress LLC Supplier No. 50MGHY78695
JQ-1 carboxylic acid is a potent BET bromodomain inhibitor, derived from (+)-JQ-1. It is utilized for synthesizing PROTACs that target the degradation of BRD4. This compound is an off-white to yellow solid with a molecular weight of 400.88 and the chemical formula C19H17ClN4O2S.
  • Potent BET bromodomain inhibitor.
  • Used to synthesize PROTAC, targeting BRD4 degradation.
  • Soluble in DMSO (100 mg/mL).
  • Downregulates PD-L1 expression on B16F10 cells in vitro.
  • Applicable in various disease areas including cancer, and signaling pathways such as epigenetics and immunology/inflammation.
  • Store powder at -20°C for 3 years or 4°C for 2 years.