Learn More
Medchemexpress LLC JQ-1 (carboxylic acid) | 202592-23-2 | 99.7% | C19H17ClN4O2S | 10 MM/1 ML
SDP
Catalog No. 5000432667
Encompass_Preferred
JQ-1 carboxylic acid is a derivative of (+)-JQ-1 and functions as a potent BET bromodomain inhibitor. It can be utilized in the synthesis of PROTAC to target the degradation of BRD4.
  • Potent BET bromodomain inhibitor
  • Used in the synthesis of PROTAC
  • Targets degradation of BRD4
  • Off-white to yellow solid appearance
  • Stable under recommended storage conditions
  • High purity (99.69%)
Catalog No. 50-004-32667 Supplier Medchemexpress LLC Supplier No. HY7869510MM/1ML
JQ-1 carboxylic acid is a derivative of (+)-JQ-1 and functions as a potent BET bromodomain inhibitor. It can be utilized in the synthesis of PROTAC to target the degradation of BRD4.
  • Potent BET bromodomain inhibitor
  • Used in the synthesis of PROTAC
  • Targets degradation of BRD4
  • Off-white to yellow solid appearance
  • Stable under recommended storage conditions
  • High purity (99.69%)