Learn More
Medchemexpress LLC HY-78695 500mg , JQ-1 (carboxylic acid) CAS:202592-23-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY78695500MG
Encompass_Preferred
HY-78695 500mg JQ-1 (carboxylic acid) CAS: 202592-23-2 JQ-1 carboxylic acid, a type of (+)-JQ-1 derivative, is a potent BET bromodomain inhibitor for downregulating PD-L1 expression on the surface of tumor cells. JQ-1 carboxylic acid can be used as a precursor to synthesize PROTACs, which targets BET bromodomains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9570