Learn More
Selleck Chemical LLC JQ-EZ-05 (JQEZ5) 2mg 1913252-04-6
SDP
Supplier:  Selleck Chemical LLC S86072MG
Encompass_Preferred
JQ-EZ-05 (JQEZ5) is a specific and reversible EZH1/2 inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8254