Learn More
Apexbio Technology LLC (-)-JQ1, 100mg. Cas: 1268524-71-5 MFCD: MFCD22124456. Stereoisomer of (+)-JQ1, used as negative control
SDP
Supplier:  Apexbio Technology LLC A8181100
Encompass_Preferred
(-)-JQ1 is the stereoisomer of JQ1, a cell-permeable small-molecule inhibitor of BET bromodomain that competitively binds to acetyl-lysine recognition motifs. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5069