Learn More
Apexbio Technology LLC (-)-JQ1 1268524-71-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061952
Encompass_Preferred
(-)-JQ1 (CAS 1268524-71-5) is a stereoisomer of ( )-JQ1 designed as a negative control in bromodomain inhibition studies Unlike ( )-JQ1 a competitive BET bromodomain inhibitor known to displace BRD4 fusion oncoproteins from chromatin the (-)-enantiomer shows negligible interaction with tested bromodomains It exhibits minimal inhibitory activity against BRD4(1) with an IC50 of 10 000 nM Due to the lack of significant bromodomain binding (-)-JQ1 serves widely as an appropriate negative control compound in experiments investigating BET protein functions and related signaling
Catalog No. 50-000-61952 Supplier Apexbio Technology LLC Supplier No. A81815.1
(-)-JQ1 (CAS 1268524-71-5) is a stereoisomer of ( )-JQ1 designed as a negative control in bromodomain inhibition studies Unlike ( )-JQ1 a competitive BET bromodomain inhibitor known to displace BRD4 fusion oncoproteins from chromatin the (-)-enantiomer shows negligible interaction with tested bromodomains It exhibits minimal inhibitory activity against BRD4(1) with an IC50 of 10 000 nM Due to the lack of significant bromodomain binding (-)-JQ1 serves widely as an appropriate negative control compound in experiments investigating BET protein functions and related signaling