Learn More
Abcam (+)-JQ1, BET bromodomain inhibitor, 1MG
Catalog No. 502706563
Encompass_Preferred
MW 457 Da, Purity >99%. Potent, selective and cell-permeable BET bromodomain inhibitor (Kd values are 50 and 90 nM at BRD4 first and second bromodomains, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6563 Supplier Abcam Supplier No. AB1414981MG
MW 457 Da, Purity >99%. Potent, selective and cell-permeable BET bromodomain inhibitor (Kd values are 50 and 90 nM at BRD4 first and second bromodomains, respectively).
The product is subject to the following: Abcam Restricted Use Statement