Learn More
Medchemexpress LLC HY-112789 10mg Medchemexpress, (+)-JQ1 PA CAS:2115701-93-2 Purity:>98%
SDP
Catalog No. 501969172
Encompass_Preferred
Medchemexpress, HY-112789 10mg (+)-JQ1 PA CAS:2115701-93-2 (+)-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1, with an IC50 of 10.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9172 Supplier Medchemexpress LLC Supplier No. HY11278910MG
Medchemexpress, HY-112789 10mg (+)-JQ1 PA CAS:2115701-93-2 (+)-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1, with an IC50 of 10.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.