Learn More
Medchemexpress LLC (+)-JQ1 PA | 2115701-93-2 | 98.2% | 437.95 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11278910MM/1ML
Encompass_Preferred
(+)-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1, with an IC50 of 10.4 nM. It is a click chemistry reagent containing an Alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Derivative of the BET inhibitor JQ1.
  • IC50 of 10.4 nM for BET.
  • Click chemistry reagent with an alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc).
Catalog No. 50-004-35773