Learn More
Selleck Chemical LLC ()-JQ1 PA S2941-5mg
SDP
Catalog No. 503111829
Encompass_Preferred
( )-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1
Catalog No. 50-311-1829 Supplier Selleck Chemical LLC Supplier No. S29415MG
Add to Cart
Add to Cart
( )-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1