Learn More
Cayman Chemical -JQ1 PosItIve Control 40nmol
SDP
Catalog No. 502896820
Encompass_Preferred
A p38a MAPK inhibitor (IC50 5500 nM in an HTRF assay) inhibits LPS-induced TNF-a production in THP-1 cells (EC50 6200 nM) has been used in the study of p38a MAPK inhibitor binding interactions and kinetics
Catalog No. 50-289-6820 Supplier Cayman Chemical Supplier No. 60050540 NMOL
A p38a MAPK inhibitor (IC50 5500 nM in an HTRF assay) inhibits LPS-induced TNF-a production in THP-1 cells (EC50 6200 nM) has been used in the study of p38a MAPK inhibitor binding interactions and kinetics