Learn More
Cayman Chemical -JQ1 PosItIve Control 8nmol
SDP
Catalog No. 502892409
Encompass_Preferred
A PEGylated derivative of DSPE has been used in CuAAC and SPAAC has been used to attach VCAM-1 antibodies to the surface of liposomes encapsulating fingolimod to provide targeted delivery of fingolimod to inflamed vasculature in a mouse model of post-stroke neuroinflammation has been used to attach FMP to the surface of synthetic platelets used in a mouse model of liver injury-induced uncontrolled hemorrhage
Catalog No. 50-289-2409 Supplier Cayman Chemical Supplier No. 6005058 NMOL
A PEGylated derivative of DSPE has been used in CuAAC and SPAAC has been used to attach VCAM-1 antibodies to the surface of liposomes encapsulating fingolimod to provide targeted delivery of fingolimod to inflamed vasculature in a mouse model of post-stroke neuroinflammation has been used to attach FMP to the surface of synthetic platelets used in a mouse model of liver injury-induced uncontrolled hemorrhage