Learn More
Cayman Chemical -JQ1free acId 10mg
SDP
Catalog No. 502895307
Encompass_Preferred
An inhibitor of BET family proteins; binds to BRD4 bromodomains 1 and 2 with Kd values of ~50 and 90 nM, respectively
Catalog No. 50-289-5307 Supplier Cayman Chemical Supplier No. 900291010 MG
An inhibitor of BET family proteins; binds to BRD4 bromodomains 1 and 2 with Kd values of ~50 and 90 nM, respectively