Learn More
Medchemexpress LLC HY-100846 5mg Medchemexpress, JQEZ5 CAS:1913252-04-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1008465MG
Encompass_Preferred
Medchemexpress, HY-100846 5mg JQEZ5 CAS:1913252-04-6 JQEZ5 is a potent and selective EZH2 lysine methyltransferase inhibitor. JQEZ5 SAM-competitive inhibition of polycomb repressive complex 2 (PRC2) with an IC50 of 80 nM. JQEZ5 has anti-tumor effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0605