Learn More
Medchemexpress LLC JQKD82 trihydrochloride | 2863676-87-1 | 99.89% | 610.01 | 50 MG
SDP
Catalog No. 5000351993
Encompass_Preferred
JQKD82 (JADA82) trihydrochloride is a cell-permeable and selective KDM5 inhibitor. It increases H3K4me3 and can be used for the research of multiple myeloma.
  • Increases global H3K4me3 levels in cells.
  • Inhibits cell growth in a dose- and time-dependent manner (IC50=0.42 μM).
  • Induces G1 cell-cycle arrest.
  • Exhibits anti-multiple myeloma activity in vivo.
  • Leads to increased H3K4me3 levels and reduced MYC immuno-staining.
  • Significantly reduces tumor burden in mouse models.
Catalog No. 50-003-51993 Supplier Medchemexpress LLC Supplier No. HY138691A50MG
JQKD82 (JADA82) trihydrochloride is a cell-permeable and selective KDM5 inhibitor. It increases H3K4me3 and can be used for the research of multiple myeloma.
  • Increases global H3K4me3 levels in cells.
  • Inhibits cell growth in a dose- and time-dependent manner (IC50=0.42 μM).
  • Induces G1 cell-cycle arrest.
  • Exhibits anti-multiple myeloma activity in vivo.
  • Leads to increased H3K4me3 levels and reduced MYC immuno-staining.
  • Significantly reduces tumor burden in mouse models.