Learn More
Medchemexpress LLC JQKD82 trihydrochloride | 2863676-87-1 | 99.9% | C27H43Cl3N4O5 | 10MG
SDP
Catalog No. 5000260895
Encompass_Preferred
JQKD82 trihydrochloride is a cell-permeable, selective inhibitor of lysine demethylase KDM5 (JARID1) used in epigenetic research. It increases histone H3 lysine 4 trimethylation (H3K4me3) and is applied in studies of multiple myeloma and KDM5-mediated regulation.
  • Selective KDM5 inhibitor.
  • Cell-permeable small molecule for intracellular studies.
  • Increases H3K4me3 levels in treated cells.
  • Supplied as the trihydrochloride salt for enhanced stability.
  • High purity suitable for research use (99.9%).
  • Available in multiple milligram pack sizes for assay flexibility.
Catalog No. 50-002-60895 Supplier Medchemexpress LLC Supplier No. HY138691A10MG
JQKD82 trihydrochloride is a cell-permeable, selective inhibitor of lysine demethylase KDM5 (JARID1) used in epigenetic research. It increases histone H3 lysine 4 trimethylation (H3K4me3) and is applied in studies of multiple myeloma and KDM5-mediated regulation.
  • Selective KDM5 inhibitor.
  • Cell-permeable small molecule for intracellular studies.
  • Increases H3K4me3 levels in treated cells.
  • Supplied as the trihydrochloride salt for enhanced stability.
  • High purity suitable for research use (99.9%).
  • Available in multiple milligram pack sizes for assay flexibility.