Learn More
Medchemexpress LLC JQKD82 trihydrochloride | 2863676-87-1 | MFCD34676876 | 99.9% | 610.01 g/mol | C27H43Cl3N4O5 | 5 MG
SDP
Catalog No. 5000221407
Encompass_Preferred
JQKD82 trihydrochloride is the trihydrochloride salt of a selective, cell-permeable small-molecule inhibitor of KDM5 histone demethylases. It elevates H3K4me3 levels in cells and is used in cellular and epigenetic studies, including multiple myeloma research. Molecular formula C27H43Cl3N4O5; molecular weight 610.01 g/mol; reported purity 99.89%.
  • Selective KDM5 inhibitor suitable for cellular assays.
  • Increases H3K4me3 levels in treated cells.
  • Cell-permeable small-molecule salt form.
  • High reported purity of 99.89%.
  • Supplied as solid and as DMSO stock solutions.
  • Useful for epigenetic and cancer research applications.
Catalog No. 50-002-21407 Supplier Medchemexpress LLC Supplier No. HY138691A5MG
JQKD82 trihydrochloride is the trihydrochloride salt of a selective, cell-permeable small-molecule inhibitor of KDM5 histone demethylases. It elevates H3K4me3 levels in cells and is used in cellular and epigenetic studies, including multiple myeloma research. Molecular formula C27H43Cl3N4O5; molecular weight 610.01 g/mol; reported purity 99.89%.
  • Selective KDM5 inhibitor suitable for cellular assays.
  • Increases H3K4me3 levels in treated cells.
  • Cell-permeable small-molecule salt form.
  • High reported purity of 99.89%.
  • Supplied as solid and as DMSO stock solutions.
  • Useful for epigenetic and cancer research applications.