Learn More
TARGETMOL CHEMICALS INC JR-AB2-011 5MG
SDP
Catalog No. 502581515
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JR-AB2-011 is a selective mTORC2 inhibitor with an IC 50 value of 0.36 uM. JR-AB2-011 inhibits mTORC2 activity by blocking Rictor-mTOR association ( K i 0.19 uM) and is cytotoxic in glioblastoma [1]. purity: 98%
Catalog No. 50-258-1515 Supplier TARGETMOL CHEMICALS INC Supplier No. T117285MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JR-AB2-011 is a selective mTORC2 inhibitor with an IC 50 value of 0.36 uM. JR-AB2-011 inhibits mTORC2 activity by blocking Rictor-mTOR association ( K i 0.19 uM) and is cytotoxic in glioblastoma [1]. purity: 98%