Learn More
Medchemexpress LLC HY-122022 5mg Medchemexpress, JR-AB2-011 CAS: Purity:>98%
SDP
Catalog No. 501932564
Encompass_Preferred
Medchemexpress, HY-122022 5mg JR-AB2-011 CAS: JR-AB2-011 is a selective mTORC2 inhibitor with an IC 50 value of 0.36 μM. JR-AB2-011 inhibits mTORC2 activity by blocking Rictor-mTOR association ( K i : 0.19 μM). JR-AB2-011 has anti-glioblastoma multiforme (GBM) properties [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2564 Supplier Medchemexpress LLC Supplier No. HY1220225MG
Medchemexpress, HY-122022 5mg JR-AB2-011 CAS: JR-AB2-011 is a selective mTORC2 inhibitor with an IC 50 value of 0.36 μM. JR-AB2-011 inhibits mTORC2 activity by blocking Rictor-mTOR association ( K i : 0.19 μM). JR-AB2-011 has anti-glioblastoma multiforme (GBM) properties [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.