Learn More
Selleck Chemical LLC JR-AB2-011-E1151-10MM/1ML
SDP
Catalog No. 503137458
Encompass_Preferred
JR-AB2-011 is a potent inhibitor of mTOR-mLST8-mSIN1-Rictor complex (mTORC2) which can block mTORC2 signaling and Rictor association with mTOR at lower effective concentrations
Catalog No. 50-313-7458 Supplier Selleck Chemical LLC Supplier No. E115110MM/1ML
Add to Cart
Add to Cart
JR-AB2-011 is a potent inhibitor of mTOR-mLST8-mSIN1-Rictor complex (mTORC2) which can block mTORC2 signaling and Rictor association with mTOR at lower effective concentrations