Learn More
Research Products International Corp UniRack Jr. Micro-Tube Rack, Blue, 10 per Case
SDP
Catalog No. 50476659
Encompass
The ultra-compact UniRackJr. provides both a convenient workstation and freezer storage rack. Use for a variety of tubes sizes such as 1.5-2.0ml tubes, 10-12 x 75mm. Turn rack over for storing 0.2-0.5ml PCR tubes. Constructed from heavy duty polypropylene for durability and chemical resistance. Autoclavable.
Catalog No. 50-476-659 Supplier Research Products International Corp Supplier No. 248162
The ultra-compact UniRackJr. provides both a convenient workstation and freezer storage rack. Use for a variety of tubes sizes such as 1.5-2.0ml tubes, 10-12 x 75mm. Turn rack over for storing 0.2-0.5ml PCR tubes. Constructed from heavy duty polypropylene for durability and chemical resistance. Autoclavable.