Learn More
Medchemexpress LLC HY-138161 5mg , JR14a CAS:2411440-41-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1381615MG
Encompass_Preferred
HY-138161 5mg JR14a CAS: 2411440-41-8 JR14a is a potent thiophene antagonist of human complement C3a receptor. JR14a shows selectivity for the human C3a receptor over C5a receptor. JR14a can suppress C3aR-mediated inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0808