Learn More
ATCC J.RT3-T3.5; Leukemia; Human (Homo sapiens)
Catalog No. 502383790
Encompass
J.RT3-T3.5 is a derivative mutant of the Jurkat leukemia cell line that lacks the beta chain of the T-cell antigen receptor. The Jurkat cell line was established from the peripheral blood of a 14-year-old, male, acute T-cell leukemia patient. This cell line can be used in immune system disorder research and immunology research.
Catalog No. 50-238-3790 Supplier ATCC Supplier No. TIB153
J.RT3-T3.5 is a derivative mutant of the Jurkat leukemia cell line that lacks the beta chain of the T-cell antigen receptor. The Jurkat cell line was established from the peripheral blood of a 14-year-old, male, acute T-cell leukemia patient. This cell line can be used in immune system disorder research and immunology research.