Learn More
Beckman Coulter Beckman Coulter SX4750 (A) Bucket Covers
Catalog No. 501631987
Encompass_Preferred
BioCertified Bucket Covers for SX4750 and SX4750A Rotors (Set of 2)
This item is not returnable. View return policy
Catalog No. 50-163-1987 Supplier Beckman Coulter Supplier No. 392805
BioCertified Bucket Covers for SX4750 and SX4750A Rotors (Set of 2)