Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc JS L80 4UM 80A 250X6MM
SDP

Catalog No. 50874482
Encompass_Preferred

Js L80 4Um; 80A 250 X 6mm

Catalog No. 50-874-482 Supplier YMC America, Inc Supplier No. JL08S042506WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Js L80 4Um; 80A 250 X 6mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.