Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc JS M80 4UM 80A 150X4.6MM
SDP

Catalog No. 50874495
Encompass_Preferred

Js M80 4Um; 80A 150 X 4.6mm

Catalog No. 50-874-495 Supplier YMC America, Inc Supplier No. JM08S041546WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Js M80 4Um; 80A 150 X 4.6mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.