Learn More
Beckman Coulter Beckman Coulter JS-4.3 Swinging-Bucket Rotor and Buckets for Avanti J-26 XP / J-HC centrifuges
Catalog No. NC3237303
Encompass
Beckman Coulter JS-4.3 swinging-bucket rotor (MPN 362734) is a four-place stainless steel rotor with anodized aluminum buckets designed for Avanti series centrifuges. It accepts modular disk adapters to accommodate tubes, bottles, and blood bag cups, and can be used with aerosol canisters for added operator protection.
  • Four-place swinging-bucket design with anodized aluminum buckets.
  • Maximum speed 4,300 rpm and maximum RCF 4,220 xg.
  • Nominal capacity approximately 3 L (4 x 750 mL).
  • Stainless steel rotor construction for durability.
  • Accommodates modular disk adapters and a wide range of tube and bottle adapters.
  • Designed for rapid sedimentation of cells, large particles, and serum separation.
  • Compatible with aerosol containment canisters for reduced operator exposure.
Catalog No. NC3237303 Supplier Beckman Coulter Supplier No. 362734
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Beckman Coulter JS-4.3 swinging-bucket rotor (MPN 362734) is a four-place stainless steel rotor with anodized aluminum buckets designed for Avanti series centrifuges. It accepts modular disk adapters to accommodate tubes, bottles, and blood bag cups, and can be used with aerosol canisters for added operator protection.
  • Four-place swinging-bucket design with anodized aluminum buckets.
  • Maximum speed 4,300 rpm and maximum RCF 4,220 xg.
  • Nominal capacity approximately 3 L (4 x 750 mL).
  • Stainless steel rotor construction for durability.
  • Accommodates modular disk adapters and a wide range of tube and bottle adapters.
  • Designed for rapid sedimentation of cells, large particles, and serum separation.
  • Compatible with aerosol containment canisters for reduced operator exposure.