Learn More
Selleck Chemical LLC JSH-150 2mg
SDP
Supplier:  Selleck Chemical LLC S87832MG
Encompass_Preferred
JSH-150 is a highly selective and potent inhibitor of CDK9 with IC50 of 1 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3391