Learn More
Medchemexpress LLC HY-X0150 5mg Medchemexpress, JSH-150 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYX01505MG
Encompass_Preferred
Medchemexpress, HY-X0150 5mg JSH-150 CAS: JSH-150 is a highly selective and potent CDK9 inhibitor with an IC 50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4666