Learn More
Medchemexpress LLC HY-13982 10mg , JSH-23 CAS:749886-87-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1398210MG
Encompass_Preferred
HY-13982 10mg JSH-23 CAS: 749886-87-1 JSH-23 is an NF-κB inhibitor which inhibits NF-κB transcriptional activity with an IC50 of 7.1 μM in lipopolysaccharide (LPS)-stimulated macrophages RAW 264.7. JSH-23 inhibits nuclear translocation of NF-κB p65 without affecting IκBα degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0839