Learn More
Apexbio Technology LLC JSH-23 749886-87-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B16455.1
Encompass_Preferred
JSH-23 (CAS 749886-87-1) is a small-molecule inhibitor of NF- B transcriptional activity exhibiting an IC50 of approximately 7 1 M Mechanistically JSH-23 prevents NF- B-mediated gene transcription by reducing nuclear localization and DNA binding activity of the NF- B p65 subunit without interfering with I B degradation In LPS-stimulated RAW 264 7 macrophages JSH-23 decreases expression of pro-inflammatory mediators including IL-6 IL-1 COX-2 and TNF- and inhibits apoptotic chromatin condensation This compound serves as a research tool for studying NF- B signaling and related inflammatory processes
Catalog No. 50-000-61738