Learn More
Selleck Chemical LLC JSH-23 5mg 749886-87-1
SDP
Supplier:  Selleck Chemical LLC S73515MG
Encompass_Preferred
JSH-23 is an inhibitor of NF-?B transcriptional activity with IC50 of 7.1 ?M in RAW 264.7 cell line. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4672