Learn More
Medchemexpress LLC HY-13982 5mg Medchemexpress, JSH-23 CAS:749886-87-1 Purity:>98%
SDP
Catalog No. 502030499
Encompass_Preferred
Medchemexpress, HY-13982 5mg JSH-23 CAS:749886-87-1 JSH-23 is an NF-κB inhibitor which inhibits NF-κB transcriptional activity with an IC50 of 7.1 μM in lipopolysaccharide (LPS)-stimulated macrophages RAW 264.7. JSH-23 inhibits nuclear translocation of NF-κB p65 without affecting IκBα degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0499 Supplier Medchemexpress LLC Supplier No. HY139825MG
Medchemexpress, HY-13982 5mg JSH-23 CAS:749886-87-1 JSH-23 is an NF-κB inhibitor which inhibits NF-κB transcriptional activity with an IC50 of 7.1 μM in lipopolysaccharide (LPS)-stimulated macrophages RAW 264.7. JSH-23 inhibits nuclear translocation of NF-κB p65 without affecting IκBα degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.