Learn More
TARGETMOL CHEMICALS INC JT010 10MG
SDP
Catalog No. 502577351
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JT010 is an effective agonist of TRPA1 (EC50 = 0.65 nM). purity: 98%
Catalog No. 50-257-7351 Supplier TARGETMOL CHEMICALS INC Supplier No. T1562810MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JT010 is an effective agonist of TRPA1 (EC50 = 0.65 nM). purity: 98%