Learn More
Medchemexpress LLC HY-111132 5mg Medchemexpress, JT010 CAS:917562-33-5 Purity:>98%
SDP
Catalog No. 501932355
Encompass_Preferred
Medchemexpress, HY-111132 5mg JT010 CAS:917562-33-5 JT010 is a potent agonist of TRPA1 with an EC 50 of 0.65 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2355 Supplier Medchemexpress LLC Supplier No. HY1111325MG
Medchemexpress, HY-111132 5mg JT010 CAS:917562-33-5 JT010 is a potent agonist of TRPA1 with an EC 50 of 0.65 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.