Learn More
Chemglass Life Sciences Thermometer, Non-Mercury, 10/18 Joint, 51mm Immersion, -10C to 250C
SDP
Catalog No. CG3508N04
Encompass_Preferred
Thermometer, Non-Mercury, 10/18 Joint, 51mm Immersion, -10C to 250C. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. Thermometer has a white back, a 6 to 7mm outer diameter, and a standard taper 10/18 inner ground joint. All ranges have 1° subdivisions. Immersion is measured from the bottom of the joint to the bottom of non-mercury bulb.
Catalog No. CG3508N04 Supplier Chemglass Life Sciences Supplier No. CG3508N04
Thermometer, Non-Mercury, 10/18 Joint, 51mm Immersion, -10C to 250C. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. Thermometer has a white back, a 6 to 7mm outer diameter, and a standard taper 10/18 inner ground joint. All ranges have 1° subdivisions. Immersion is measured from the bottom of the joint to the bottom of non-mercury bulb.