Learn More
Medchemexpress LLC HY-100675 5mg Medchemexpress, JTE-013 CAS:383150-41-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006755MG
Encompass_Preferred
Medchemexpress, HY-100675 5mg JTE-013 CAS:383150-41-2 JTE-013 is a potent and specific S1P2 (Sphingosine-1-Phosphate 2; EDG-5) antagonist. JTE-013 inhibits the specific binding of radiolabeled S1P to human and rat S1P2 with IC50s of 17 nM and 22 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7765