Learn More
Medchemexpress LLC HY-110133 5mg , JTE-607 CAS:188791-09-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1101335MG
Encompass_Preferred
HY-110133 5mg JTE-607 CAS: 188791-09-5 JTE-607, a highly selective inflammatory cytokine synthesis inhibitor, protects from endotoxin shock in mice. JTE-607 inhibits inflammatory cytokine production, including TNF-α, IL-1β, IL-6, IL-8 and IL-10, from LPS-stimulated human PBMCs, with IC50s of 11, 5.9, 8.8, 7.3 and 9.1 nM, respectively. Cleavage and Polyadenylation Specificity Factor 3 (CPSF3) is the target of JTE-607. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9933