Learn More
Abcam JTE 607 dihydrochloride, 25MG
Catalog No. 502682955
Encompass_Preferred
MW 597.4 Da, Purity >98%. Inhibits inflammatory cytokine production in human peripheral blood mononuclear cells (PBMC's), e.g. IC₅₀'s = 11 nM (TNF-α), 5.9 nM (IL-1β), 8.8 nM (IL-6), 7.3 nM (IL-8), and 9.1 nM (IL-10). Induces apoptosis in leukemia cells in vitro and prolongs survival in a mouse leukemia model.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2955 Supplier Abcam Supplier No. AB27339025MG
MW 597.4 Da, Purity >98%. Inhibits inflammatory cytokine production in human peripheral blood mononuclear cells (PBMC's), e.g. IC₅₀'s = 11 nM (TNF-α), 5.9 nM (IL-1β), 8.8 nM (IL-6), 7.3 nM (IL-8), and 9.1 nM (IL-10). Induces apoptosis in leukemia cells in vitro and prolongs survival in a mouse leukemia model.
The product is subject to the following: Abcam Restricted Use Statement