Learn More
Selleck Chemical LLC JTE-607 Dihydrochloride
SDP
Supplier:  Selleck Chemical LLC E0314100MG
Encompass_Preferred
JTE-607 Dihydrochloride is a prodrug compound that is converted to an active form by ester hydrolysis JTE-607 is a multiple cytokine production inhibitor that targets CPSF3 and inhibits pre-mRNA processing JTE-607 inhibits tumor necrosis factor (TNF)-alpha interleukin (IL)-1beta IL-6 IL-8 and IL-10 from LPS-stimulated human PBMCs with IC50 values of 11 nM 5 9 nM 8 8 nM 7 3 nM and 9 1 nM respectively
Catalog No. 50-313-6801