Learn More
Research Products International Corp JTR Press-to-Seal Silicone Isolator, 2.0mm Thick, 2.5mm Diameter, 25/pk
SDP
Catalog No. 501977497
Encompass
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Nonadhesive isolators are autoclavable and reusable.
Catalog No. 50-197-7497 Supplier Research Products International Corp Supplier No. 141306
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Nonadhesive isolators are autoclavable and reusable.