Learn More
TARGETMOL CHEMICALS INC JUGLONE 50MG
SDP
Catalog No. 502576832
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Juglone (Regianin) is a natural naphthoquinone found in the black walnut (J. nigra) and other plants in the Juglandaceae family. Juglone also irreversibly inhibits peptidyl-prolyl cis/trans isomerases of the parvulin family including human Pin1 yeast Ess1/Ptf1 and E. coli parvulin (Ki = 55.9 nM). Juglone also blocks transcription by RNA polymerases I II and III (IC50s = 2-7 uM) and attenuates kidney fibrosis in rats treated with unilateral ureteral obstruction both through Pin1-independent mechanisms. purity: 98%
Catalog No. 50-257-6832 Supplier TARGETMOL CHEMICALS INC Supplier No. T401050MG
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Juglone (Regianin) is a natural naphthoquinone found in the black walnut (J. nigra) and other plants in the Juglandaceae family. Juglone also irreversibly inhibits peptidyl-prolyl cis/trans isomerases of the parvulin family including human Pin1 yeast Ess1/Ptf1 and E. coli parvulin (Ki = 55.9 nM). Juglone also blocks transcription by RNA polymerases I II and III (IC50s = 2-7 uM) and attenuates kidney fibrosis in rats treated with unilateral ureteral obstruction both through Pin1-independent mechanisms. purity: 98%