Learn More
TARGETMOL CHEMICALS INC JUJUBOSIDE B 10MG
SDP
Catalog No. 502576875
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Jujuboside B has potent inhibitory effects on collagen- thrombin- AA- and ADP-induced aggregation and also exhibits superior protection on the thromboembolic model. purity: 98%
Catalog No. 50-257-6875 Supplier TARGETMOL CHEMICALS INC Supplier No. T378110MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Jujuboside B has potent inhibitory effects on collagen- thrombin- AA- and ADP-induced aggregation and also exhibits superior protection on the thromboembolic model. purity: 98%